4-Benzyloxy-3,5-dimethylbenzoic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L14536-03
L14536-06
CAAAAL14536-03EA
97.33
CAD
CAAAAL14536-03
CAAAAL14536-06
4-Benzyloxy-3,5-dimethylbenzoic acid 98+%
4-Benzyloxy-3,5-dimethylbenzoic acid
Purity: 98+%
CAS No.: 97888-80-7
Molecular Formula: C16H16O3
Formula Weight: 256.30
Melting Point: 157-160°
Beilstein Reference: 5045861
Formula:
C₁₆H₁₆O₃ Melting Pt: 157…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00272572 CAS Number: 97888-80-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...