Methyl indole-3-carboxylate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L14516-14
L14516-22
CAAAAL14516-14EA
79.63
CAD
CAAAAL14516-14
CAAAAL14516-22
Methyl indole-3-carboxylate 99%
Methyl indole-3-carboxylate
Purity: 99%
CAS No.: 942-24-5
Molecular Formula: C10H9NO2
Formula Weight: 175.19
Melting Point: 149-152°
Beilstein Reference: 142023
Formula:
C₁₀H₉NO₂ Melting Pt: 149…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD00189407 CAS Number: 942-24-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...