5-Amino-2-chlorobenzoic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L14501-14
L14501-22
CAAAAL14501-14EA
79.63
CAD
CAAAAL14501-14
CAAAAL14501-22
5-Amino-2-chlorobenzoic acid 98+%
5-Amino-2-chlorobenzoic acid
Purity: 98+%
CAS No.: 89-54-3
Molecular Formula: C7H6ClNO2
Formula Weight: 171.58
Melting Point: ca 185° dec.
Beilstein Reference: 2090422
Formula:
C₇H₆ClNO₂ MW: 171.58 g/mol Boiling Pt: 330 Melting Pt: ∼ 185 °C (dec.) Density: 1.5 Storage Temperature: Ambient |
MDL Number:
MFCD00007845 CAS Number: 89-54-3 EINECS: 201-916-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...