L(+)-Sodium lactate 98+%

Supplier: Thermo Scientific Chemicals

L14500-06 L14500-14
CAAAAL14500-06EA 107.44 CAD
CAAAAL14500-06 CAAAAL14500-14
L(+)-Sodium lactate 98+%
L(+)-Sodium lactate

Purity: 98+% CAS No.: 867-56-1 Molecular Formula: C3H5NaO3 Formula Weight: 112.06 Melting Point: ca 160° Beilstein Reference: 4567087

Formula: C₃H₅NaO₃
Melting Pt: ∼ 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00066576
CAS Number: 867-56-1
EINECS: 212-762-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR