Dibutyltin oxide 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Dibutyltin(IV) oxide, Dibutyloxostannane, Di-n-butyltin oxide, Di-n-butyltin(IV) oxide

L14491-22 L14491-36
CAAAAL14491-22EA 57.67 CAD
CAAAAL14491-22 CAAAAL14491-36
Dibutyltin oxide 98%
Dibutyltin oxide

Purity: 98% CAS No.: 818-08-6 Molecular Formula: C8H18OSn Formula Weight: 248.94 Odor: Odorless Melting Point: 300° Beilstein Reference: 4126243 UN No.: 3146 MDL No.: MFCD00001992

Formula: C₈H₁₈OSn
MW: 248.94 g/mol
Melting Pt: >300 °C
Density: 1.58
Storage Temperature: Ambient
MDL Number: MFCD00001992
CAS Number: 818-08-6
EINECS: 212-449-1
UN: 3146
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR