Triethylsilyl trifluoromethanesulfonate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L14477-06
L14477-14
CAAAAL14477-06EA
84.37
CAD
CAAAAL14477-06
CAAAAL14477-14
Triethylsilyl trifluoromethanesulfonate 98%
Triethylsilyl trifluoromethanesulphonate
Purity: 98%
CAS No.: 79271-56-0
Molecular Formula: C7F3H15O3SSi
Formula Weight: 264.33
Storage Temperature: 2°C to 8°C
Flash Point: 72°(161°F)
Refractive Index: 1.3910
Boiling Point: 207°
Beilstein Reference: 3590541
UN No.: 3265
MDL No.: MFCD00000407
Formula:
C₇H₁₅F₃O₃SSi Boiling Pt: 207 °C Density: 1.225 Flash Pt: 72 °C (161 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000407 CAS Number: 79271-56-0 EINECS: 279-124-4 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...