Trichloroethylene 98%

Supplier: Thermo Scientific Chemicals
Danger

L14474-0F L14474-AE L14474-AU
CAAAAL14474-0FEA 119.29 CAD
CAAAAL14474-0F CAAAAL14474-AE CAAAAL14474-AU
Trichloroethylene 98%
Trichloroethylene

Purity: 98% CAS No.: 79-01-6 Molecular Formula: ClCH=CCl2 Formula Weight: 131.39 Flash Point: None Refractive Index: 1.476 Odor: Chloroform-like Melting Point: -85° Boiling Point: 87° Beilstein Reference: 1736782 Merck Reference: 14,9639 UN No.: 1710 MDL No.: MFCD00000838

Formula: ClCH=CCl₂
Boiling Pt: 87 °C
Melting Pt: –85 °C
Density: 1.46
Storage Temperature: Ambient
MDL Number: MFCD00000838
CAS Number: 79-01-6
EINECS: 201-167-4
UN: 1710
ADR: 6.1,III
Merck Index: 14,9639
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ac57d7a9ea2e3097408c3559993eb33587aec09&vid=767d7d09973e747c5a2d07a4d46dffedfb559ceb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR