(±)-δ-Dodecalactone 98%
Supplier: Thermo Scientific Chemicals
L14406-14
L14406-22
CAAAAL14406-14EA
54.51
CAD
CAAAAL14406-14
CAAAAL14406-22
(±)-δ-Dodecalactone 98%
(±)-δ-Dodecalactone
Purity: 98%
CAS No.: 713-95-1
Molecular Formula: C12H22O2
Formula Weight: 198.30
Flash Point: 165°(329°F)
Refractive Index: 1.4600
Melting Point: -7°
Boiling Point: 288°
Beilstein Reference: 1282749
MDL No.: MFCD00006651
Formula:
C₁₂H₂₂O₂ Boiling Pt: 288 °C Melting Pt: –7 °C Density: 0.950 Flash Pt: 165 °C(329 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006651 CAS Number: 713-95-1 EINECS: 211-932-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...