(±)-δ-Octalactone 97%

Supplier: Thermo Scientific Chemicals

Synonyms: (±)-Octan-5-olide, (±)-Tetrahydro-6-propyl-2H-pyran-2-one, δ-Octanolactone

L14303-14 L14303-22
CAAAAL14303-14EA 98.59 CAD
CAAAAL14303-14 CAAAAL14303-22
(±)-δ-Octalactone 97%
(±)-δ-Octalactone

Purity: 97% CAS No.: 698-76-0 Molecular Formula: C8H14O2 Formula Weight: 142.20 Flash Point: 125°(257°F) Refractive Index: 1.4550 Melting Point: -14° Boiling Point: 238° Beilstein Reference: 111515 MDL No.: MFCD00144051

Formula: C₈H₁₄O₂
MW: 142.2 g/mol
Boiling Pt: 238 °C
Melting Pt: –14 °C
Density: 1.002
Flash Pt: 125 °C(257 °F)
Storage Temperature: Ambient
MDL Number: MFCD00144051
CAS Number: 698-76-0
EINECS: 211-820-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR