(S)-(+)-Epichlorhydrin 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(S)-(+)-Glycerol epichlorhydrin, (S)-(+)-3-Chloro-1,2-epoxypropane, (S)-(+)-1-Chloro-2,3-epoxypropane, (2S)-2-(chloromethyl)oxirane
L14298-03
L14298-06
L14298-14
CAAAAL14298-03EA
96.22
CAD
CAAAAL14298-03
CAAAAL14298-06
CAAAAL14298-14
(S)-(+)-Epichlorhydrin 98+%
(S)-(+)-Epichlorhydrin
Purity: 98+%
CAS No.: 67843-74-7
Molecular Formula: C3H5ClO
Formula Weight: 92.53
Storage Temperature: 2°C to 8°C
Flash Point: 32°(90°F)
Refractive Index: 1.4380
Boiling Point: 116-117°
Beilstein Reference: 1420784
Optical Rotation: +35° (c=1 in methanol)
UN No.: 2023
MDL No.: MFCD00077760
Formula:
ClCH₂CH(O)CH₂ MW: 92.52 g/mol Boiling Pt: 116…117 °C Melting Pt: -48 Density: 1.180 Flash Pt: 32 °C (90 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00077760 CAS Number: 67843-74-7 UN: 2023 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...