Perchloroethane 99%

Supplier: Thermo Scientific Chemicals
Warning

L14297-22 L14297-36
CAAAAL14297-22EA 51.03 CAD
CAAAAL14297-22 CAAAAL14297-36
Perchloroethane 99%
Perchloroethane

Purity: 99% CAS No.: 67-72-1 Molecular Formula: Cl3CCCl3 Formula Weight: 236.74 Odor: Camphor-like Melting Point: 185-190° subl. Beilstein Reference: 1740341 Merck Reference: 14,4679 UN No.: 3077 MDL No.: MFCD00000799

Formula: C₂Cl₆
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: 185…190 °C (subl.)
Density: 2.091
Storage Temperature: Ambient
MDL Number: MFCD00000799
CAS Number: 67-72-1
EINECS: 200-666-4
UN: 3077
ADR: 9,III
Merck Index: 14,4679

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR