1,2-Diethoxyethane 98%

Supplier: Thermo Scientific Chemicals
Danger

L14283-AE L14283-AP
CAAAAL14283-AEEA 87.85 CAD
CAAAAL14283-AE CAAAAL14283-AP
1,2-Diethoxyethane 98%
1,2-Diethoxyethane

Purity: 98% CAS No.: 629-14-1 Molecular Formula: C6H14O2 Formula Weight: 118.18 Flash Point: 19°(66°F) Refractive Index: 1.3920 Melting Point: -74° Boiling Point: 120-121° Beilstein Reference: 1732917 UN No.: 1153 MDL No.: MFCD00009253

Formula: C₆H₁₄O₂
Boiling Pt: 120…121 °C
Melting Pt: –74 °C
Density: 0.842
Flash Pt: 19 °C(66 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009253
CAS Number: 629-14-1
EINECS: 211-076-1
UN: 1153
ADR: "3",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR