D-(-)-N-Methylglucamine 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Meglumine, (-)-1-Deoxy-1-(methylamino)-D-glucitol, N-Methyl-D-glucamine, (-)-N-Methyl-D-glucamine, (2R,3R,4R,5S)-6-(Methylamino)hexane-1,2,3,4,5-pentaol, (-)-1-Methylamino-1-deoxy-D-glucitol
L14282-18
L14282-30
CAAAAL14282-18EA
43.61
CAD
CAAAAL14282-18
CAAAAL14282-30
D-(-)-N-Methylglucamine 99%
D(-)-N-Methylglucamine
Purity: 99%
CAS No.: 6284-40-8
Molecular Formula: C7H17NO5
Formula Weight: 195.22
Melting Point: 128-131°
Beilstein Reference: 385906
Merck Reference: 14,6078
Optical Rotation: -16.5° (c=2 in water)
MDL No.: MFCD00004707
Formula:
H₃CNHCH₂(CH(OH))₄CH₂OH Melting Pt: 128…131 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004707 CAS Number: 6284-40-8 EINECS: 228-506-9 Merck Index: 14,6078 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...