2-Aminoanthracene 94%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L14269-03
L14269-06
CAAAAL14269-03EA
63.04
CAD
CAAAAL14269-03
CAAAAL14269-06
2-Aminoanthracene 94%
2-Aminoanthracene
Purity: 94%
CAS No.: 613-13-8
Molecular Formula: C14H11N
Formula Weight: 193.25
Melting Point: 235-239°
Beilstein Reference: 2209414
Formula:
C₁₄H₁₁N Boiling Pt: 371 °C (1013 hPa) Melting Pt: 235…239 °C Density: Min. 1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003582 CAS Number: 613-13-8 EINECS: 210-330-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...