2-Bromopropene 99% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
L14229-06
L14229-14
L14229-22
CAAAAL14229-06EA
87.85
CAD
CAAAAL14229-06
CAAAAL14229-14
CAAAAL14229-22
2-Bromopropene 99% stabilized
2-Bromopropene
Purity: 99%
CAS No.: 557-93-7
Molecular Formula: C3H5Br
Formula Weight: 120.98
Storage Temperature: 2°C to 8°C
Flash Point: 4°(39°F)
Refractive Index: 1.4430
Odor: Pungent
Melting Point: -87°
Boiling Point: 47-49°
Beilstein Reference: 1731926
UN No.: 1993
MDL No.: MFCD00000140
Formula:
C₃H₅Br Boiling Pt: 47…49 °C Melting Pt: –87 °C Density: 1.362 Flash Pt: 4 °C (39 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000140 CAS Number: 557-93-7 EINECS: 209-185-4 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...