(R)-(-)-Epichlorhydrin 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(R)-(-)-Glycerol epichlorhydrin, (R)-(-)-3-Chloro-1,2-epoxypropane, (R)-(-)-1-Chloro-2,3-epoxypropane
L14198-03
L14198-06
L14198-14
CAAAAL14198-03EA
88.95
CAD
CAAAAL14198-03
CAAAAL14198-06
CAAAAL14198-14
(R)-(-)-Epichlorhydrin 98+%
(R)-(-)-Epichlorhydrin
Purity: 98+%
CAS No.: 51594-55-9
Molecular Formula: C3H5ClO
Formula Weight: 92.53
Storage Temperature: 2°C to 8°C
Flash Point: 32°(90°F)
Refractive Index: 1.4380
Boiling Point: 116-117°
Beilstein Reference: 1420785
Optical Rotation: -35° (c=1 in methanol)
UN No.: 2023
MDL No.: MFCD00077759
Formula:
ClCH₂CH(O)CH₂ MW: 92.52 g/mol Boiling Pt: 116…117 °C Melting Pt: –48 °C Density: 1.180 Flash Pt: 32 °C (90 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00077759 CAS Number: 51594-55-9 EINECS: 424-280-2 UN: 2023 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...