1-Chloroethyl chloroformate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Chloroethyl carbonochloridate

L14197-06 L14197-14
CAAAAL14197-06EA 51.03 CAD
CAAAAL14197-06 CAAAAL14197-14
1-Chloroethyl chloroformate 98%
1-Chloroethyl chloroformate

Purity: 98% CAS No.: 50893-53-3 Molecular Formula: C3H4Cl2O2 Formula Weight: 142.97 Storage Temperature: 2°C to 8°C Flash Point: 45°(113°F) Refractive Index: 1.4230 Melting Point: -65° Boiling Point: 119-121° Beilstein Reference: 1747601 UN No.: 2742 MDL No.: MFCD00000647

Formula: C₃H₄Cl₂O₂
Boiling Pt: 119…121 °C
Melting Pt: –65 °C
Density: 1.312
Flash Pt: 45 °C (113 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000647
CAS Number: 50893-53-3
EINECS: 256-834-2
UN: 2742
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR