Luteolin 97%

Supplier: Thermo Scientific Chemicals
Warning

L14186-MC L14186-ME
CAAAAL14186-MCEA 121.5 CAD
CAAAAL14186-MC CAAAAL14186-ME
Luteolin 97%
Luteolin

An apoptosis inducer


Purity: 97% CAS No.: 491-70-3 Molecular Formula: C15H10O6 Formula Weight: 286.24 Melting Point: 300° Beilstein Reference: 292084 Merck Reference: 14,5614 MDL No.: MFCD00017309

Formula: C₁₅H₁₀O₆
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00017309
CAS Number: 491-70-3
EINECS: 207-741-0
Merck Index: 14,5614

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR