Baicalein 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L14185-MC
L14185-ME
CAAAAL14185-MCEA
67.78
CAD
CAAAAL14185-MC
CAAAAL14185-ME
Baicalein 97%
Baicalein
Purity: 97%
CAS No.: 491-67-8
Molecular Formula: C15H10O5
Formula Weight: 270.24
Melting Point: 268-270°
Beilstein Reference: 272683
Merck Reference: 14,942
MDL No.: MFCD00017459
Formula:
C₁₅H₁₀O₅ Melting Pt: 268…270 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017459 CAS Number: 491-67-8 Merck Index: 14,942 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...