4-(Dimethylamino)benzoyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L14176-06
L14176-14
CAAAAL14176-06EA
79.63
CAD
CAAAAL14176-06
CAAAAL14176-14
4-(Dimethylamino)benzoyl chloride 97%
4-(Dimethylamino)benzoyl chloride
Purity: 97%
CAS No.: 4755-50-4
Molecular Formula: C9H10ClNO
Formula Weight: 183.64
Melting Point: 142-148°
Beilstein Reference: 1100858
UN No.: 3261
MDL No.: MFCD00236039
Formula:
C₉H₁₀ClNO Boiling Pt: 180 °C (15 -rr) Melting Pt: 142…148 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236039 CAS Number: 4755-50-4 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...