(S)-(+)-2-Butanol 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(S)-(+)-sec-Butyl alcohol, (S)-Butan-2-ol, (S)-(+)-sec-Butanol, (S)-(+)-Ethyl methyl carbinol, (S)-(+)-2-Hydroxy butane, (S)-(+)-Methyl ethyl carbinol
For synthesis of optically active products
Purity: 98+%
CAS No.: 4221-99-2
Molecular Formula: C4H10O
Formula Weight: 74.12
Flash Point: 26°(79°F)
Refractive Index: 1.3980
Odor: Alcohol-like
Boiling Point: 99-100°
Beilstein Reference: 1718763
Merck Reference: 14,1541
Optical Rotation: +13.5° (c=10 in methanol)
UN No.: 1120
MDL No.: MFCD00064281
Formula:
C₄H₁₀O MW: 74.12 g/mol Boiling Pt: 99…100 °C Melting Pt: –114 °C Density: 0.806 Flash Pt: 26 °C (79 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064281 CAS Number: 4221-99-2 EINECS: 224-168-1 UN: 1120 ADR: 3,III Merck Index: 14,1541 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...