3-Chlorobutanone 96%

Supplier: Thermo Scientific Chemicals
Warning

L14158-22 L14158-36
CAAAAL14158-22EA 71.89 CAD
CAAAAL14158-22 CAAAAL14158-36
3-Chlorobutanone 96%
3-Chlorobutanone

Purity: 96% CAS No.: 4091-39-8 Molecular Formula: C4H7ClO Formula Weight: 106.55 Flash Point: 28°(82°F) Refractive Index: 1.4210 Melting Point: -60° Boiling Point: 113-116° Beilstein Reference: 385637 UN No.: 1224 MDL No.: MFCD00000865

Formula: C₄H₇ClO
Boiling Pt: 113…116 °C
Melting Pt: < –60 °C
Density: 1.055
Flash Pt: 28 °C(82 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000865
CAS Number: 4091-39-8
EINECS: 223-834-9
UN: 1224
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR