2,6-Di-tert-butyl-4-methylpyridine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L14143-03
L14143-06
CAAAAL14143-03EA
68.1
CAD
CAAAAL14143-03
CAAAAL14143-06
2,6-Di-tert-butyl-4-methylpyridine 97%
2,6-Di-tert-butyl-4-methylpyridine
Purity: 97%
CAS No.: 38222-83-2
Molecular Formula: C14H23N
Formula Weight: 205.35
Storage Temperature: 2°C to 8°C
Flash Point: 83°(181°F)
Refractive Index: 1.4765
Melting Point: 30-35°
Boiling Point: 232-234°
Beilstein Reference: 130503
MDL No.: MFCD00006305
Formula:
C₁₄H₂₃N Boiling Pt: 232…234 °C Melting Pt: 30…35 °C Flash Pt: 83 °C (181 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006305 CAS Number: 38222-83-2 EINECS: 253-834-4 UN: 1325 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...