4-Aminobenzamidine dihydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Aminophenylamidine dihydrochloride, 4-Aminobenzene-1-carboximidamide dihydrochloride

L14089-03 L14089-06
CAAAAL14089-03EA 66.2 CAD
CAAAAL14089-03 CAAAAL14089-06
4-Aminobenzamidine dihydrochloride 97%
4-Aminobenzamidine dihydrochloride

Purity: 97% CAS No.: 2498-50-2 Molecular Formula: C7H9N3.2HCl Formula Weight: 208.09 Storage Temperature: 2°C to 8°C Melting Point: 300° Beilstein Reference: 3692927 MDL No.: MFCD00013001

Formula: C₇H₁₁Cl₂N₃
MW: 208.09 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00013001
CAS Number: 2498-50-2
EINECS: 219-692-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR