Tetrapentylammonium iodide 98%

Supplier: Thermo Scientific Chemicals
Warning

L14088-14 L14088-22
CAAAAL14088-06EA 111.82 CAD
CAAAAL14088-06 CAAAAL14088-14 CAAAAL14088-22
Tetrapentylammonium iodide 98%
Tetrapentylammonium iodide

Purity: 98% CAS No.: 2498-20-6 Molecular Formula: C20H44IN Formula Weight: 425.49 Melting Point: 135-139° Beilstein Reference: 3917412

Formula: C₂₀H₄₄IN
Melting Pt: 135…139 °C
Density: 1.18 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041980
CAS Number: 2498-20-6
EINECS: 219-688-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR