(1S,2S)-(+)-1,2-Diaminocyclohexane 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L14072-03
L14072-06
CAAAAL14072-03EA
145.68
CAD
CAAAAL14072-03
CAAAAL14072-06
CAAAAL14072-MD
(1S,2S)-(+)-1,2-Diaminocyclohexane 98%
(1S,2S)-(+)-1,2-Diaminocyclohexane
For synthesis of optically active products
Purity: 98%
CAS No.: 21436-03-3
Molecular Formula: C6H14N2
Formula Weight: 114.19
Storage Temperature: 2°C to 8°C
Flash Point: 70°(158°F)
Melting Point: 43-45°
Boiling Point: 104-110°/40mm
Beilstein Reference: 2801645
Optical Rotation: +25.5° (c=5 in 1N HCl)
UN No.: 3259
MDL No.: MFCD00062986
Formula:
C₆H₁₄N₂ Boiling Pt: 104…110 °C/40 mm Melting Pt: 43…45 °C Flash Pt: 70 °C (158 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00062986 CAS Number: 21436-03-3 UN: 3259 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...