Tetrafluoroboric acid ∼50% (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

L14037-0E L14037-22 L14037-36
CAAAAL14037-0EEA 360.24 CAD
CAAAAL14037-0E CAAAAL14037-22 CAAAAL14037-36
Tetrafluoroboric acid ∼50% (w/w) in aqueous solution
Tetrafluoroboric acid

CAS No.: 16872-11-0 Molecular Formula: HBF4 Formula Weight: 87.81 Odor: Pungent Boiling Point: 130° dec. Merck Reference: 14,4149 MDL No.: MFCD00011345

Formula: HBF₄
Boiling Pt: 130 °C (dec.)
Density: 1.4
Storage Temperature: Ambient
MDL Number: MFCD00011345
CAS Number: 16872-11-0
EINECS: 240-898-3
UN: 1775
ADR: 8,II
Merck Index: 14,4149

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR