Acetyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Acetyl chloride 1M in Dichloromethane
L14013-0F
L14013-AC
L14013-AP
CAAAAL14013-0FEA
297.04
CAD
CAAAAL14013-0F
CAAAAL14013-AC
CAAAAL14013-AP
Acetyl chloride 98%
Acetyl chloride
Purity: 98%
CAS No.: 75-36-5
Molecular Formula: C2H3ClO
Formula Weight: 78.50
Flash Point: 4°(39°F)
Refractive Index: 1.3890
Odor: Pungent
Melting Point: -112°
Boiling Point: 50-52°
Beilstein Reference: 605303
Merck Reference: 14,85
UN No.: 1717
MDL No.: MFCD00000719
Formula:
C₂H₃ClO MW: 78.5 g/mol Boiling Pt: 50.9 Melting Pt: -112 Density: 1.104 Flash Pt: 4 °C (39 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000719 CAS Number: 75-36-5 EINECS: 200-865-6 UN: 1717 ADR: 3 II Merck Index: 14,85 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...