Silica gel, Pore size: 60 Å 0.036-0.071 mm (215-400 mesh)

Supplier: Thermo Scientific Chemicals

L14004-0B L14004-0E L14004-36
CAAAAL14004-0BEA 167.48 CAD
CAAAAL14004-0B CAAAAL14004-0E CAAAAL14004-36
Silica gel, Pore size: 60 Å 0.036-0.071 mm (215-400 mesh)
Silica gel

CAS No.: 7631-86-9 Odor: Odorless Merck Reference: 14,8493 MDL No.: MFCD00011232

Formula: SiO₂
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
EINECS: 231-545-4
Merck Index: 14,8493

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR