(S)-(-)-2-Phenyl-1-propanol 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L13988-03
L13988-MD
CAAAAL13988-03EA
301.78
CAD
CAAAAL13988-03
CAAAAL13988-MD
(S)-(-)-2-Phenyl-1-propanol 98+%
(S)-(-)-2-Phenyl-1-propanol
For synthesis of optically active products
Purity: 98+%
CAS No.: 37778-99-7
Molecular Formula: C9H12O
Formula Weight: 136.19
Flash Point: 108°(226°F)
Refractive Index: 1.5260
Boiling Point: 220°
Optical Rotation: -18° (neat)
MDL No.: MFCD00145249
Formula:
C₉H₁₂O Boiling Pt: 220 °C Density: 0.975 Flash Pt: 108 °C(226 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00145249 CAS Number: 37778-99-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...