(S)-(+)-2-Aminobutyric acid 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Abu-OH, L-alpha-Amino-N-Butyric acid, L-2-Aminobutyric acid, (S)-2-Aminobutanoic acid

L13984-03 L13984-06 L13984-14
CAAAAL13984-03EA 96.06 CAD
CAAAAL13984-03 CAAAAL13984-06 CAAAAL13984-14
(S)-(+)-2-Aminobutyric acid 98+%
(S)-(+)-2-Aminobutyric acid

Purity: 98+% CAS No.: 1492-24-6 Molecular Formula: C4H9NO2 Formula Weight: 103.12 Melting Point: 300° Beilstein Reference: 1720935 Merck Reference: 14,428 Optical Rotation: +21° (c=4.8 in 6N HCl) MDL No.: MFCD00064415

Formula: C₄H₉NO₂
MW: 103.12 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00064415
CAS Number: 1492-24-6
EINECS: 216-083-3
Merck Index: 14,428

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR