(R)-(-)-2-Butanol 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L13983-03
L13983-06
CAAAAL13983-03EA
106.33
CAD
CAAAAL13983-03
CAAAAL13983-06
(R)-(-)-2-Butanol 98+%
(R)-(-)-2-Butanol
For synthesis of optically active products
Purity: 98+%
CAS No.: 14898-79-4
Molecular Formula: C4H10O
Formula Weight: 74.12
Flash Point: 26°(79°F)
Refractive Index: 1.3980
Odor: Alcohol-like
Boiling Point: 99-100°
Beilstein Reference: 1718764
Merck Reference: 14,1541
Optical Rotation: -15° (c=10 in methanol)
UN No.: 1120
MDL No.: MFCD00064280
Formula:
C₄H₁₀O Boiling Pt: 99…100 °C Melting Pt: –114 °C Density: 0.806 Flash Pt: 26 °C (79 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064280 CAS Number: 14898-79-4 EINECS: 238-967-8 UN: 1120 ADR: 3,III Merck Index: 14,1541 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...