6-Aminoquinoline 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Quinolin-6-ylamine

L13819-03 L13819-06 L13819-14
CAAAAL13819-03EA 75.84 CAD
CAAAAL13819-03 CAAAAL13819-06 CAAAAL13819-14
6-Aminoquinoline 98%
6-Aminoquinoline

Purity: 98% CAS No.: 580-15-4 Molecular Formula: C9H8N2 Formula Weight: 144.18 Odor: Odorless Melting Point: 115-119° Boiling Point: 146°/0.3mm Beilstein Reference: 113320 MDL No.: MFCD00006803

Formula: C₉H₈N₂
MW: 144.18 g/mol
Boiling Pt: 376
Melting Pt: 115…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00006803
CAS Number: 580-15-4
EINECS: 209-453-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR