3-Quinolinecarbaldehyde 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L13815-03
L13815-06
L13815-MD
CAAAAL13815-03EA
183.28
CAD
CAAAAL13815-03
CAAAAL13815-06
CAAAAL13815-MD
3-Quinolinecarbaldehyde 98+%
3-Quinolinecarbaldehyde
Purity: 98+%
CAS No.: 13669-42-6
Molecular Formula: C10H7NO
Formula Weight: 157.17
Melting Point: 68-71°
Beilstein Reference: 113269
Formula:
C₁₀H₇NO Melting Pt: 68…71 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006768 CAS Number: 13669-42-6 EINECS: 237-147-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...