(2,4-Dimethoxyphenyl)acetic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,4-Dimethoxyphenylacetic Acid
L13784-03
L13784-06
CAAAAL13784-03EA
45.98
CAD
CAAAAL13784-03
CAAAAL13784-06
(2,4-Dimethoxyphenyl)acetic acid 98%
(2,4-Dimethoxyphenyl)acetic acid
Purity: 98%
CAS No.: 6496-89-5
Molecular Formula: C10H12O4
Formula Weight: 196.20
Melting Point: 108-112°
Beilstein Reference: 2694132
Formula:
(C₆H₃(OCH₃)₂)CH₂COOH MW: 196.2 g/mol Melting Pt: 108…112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00183328 CAS Number: 6496-89-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...