4-(2-Hydroxyethyl)pyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L13780-03
L13780-06
CAAAAL13780-03EA
109.34
CAD
CAAAAL13780-03
CAAAAL13780-06
4-(2-Hydroxyethyl)pyridine 98%
4-(2-Hydroxyethyl)pyridine
Purity: 98%
CAS No.: 5344-27-4
Molecular Formula: C7H9NO
Formula Weight: 123.16
Flash Point: 92°(197°F)
Refractive Index: 1.5380
Melting Point: 8-10°
Boiling Point: 121-122°/2mm
Beilstein Reference: 111361
MDL No.: MFCD00129038
Formula:
C₇H₉NO Boiling Pt: 121…122 °C/2 mm Melting Pt: 8…10 °C Density: 1.093 Flash Pt: 92 °C(197 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00129038 CAS Number: 5344-27-4 EINECS: 226-286-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...