3,4-Dibromobenzaldehyde 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L13762-03
L13762-06
CAAAAL13762-03EA
145.36
CAD
CAAAAL13762-03
CAAAAL13762-06
3,4-Dibromobenzaldehyde 99%
3,4-Dibromobenzaldehyde
Purity: 99%
CAS No.: 74003-55-7
Molecular Formula: C7H4Br2O
Formula Weight: 263.93
Melting Point: 69-72°
Beilstein Reference: 2573433
Formula:
C₇H₄Br₂O Melting Pt: 69…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236033 CAS Number: 74003-55-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...