Isophorone diisocyanate (mixture of isomers) 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L13759-06
L13759-22
L13759-36
CAAAAL13759-06EA
47.87
CAD
CAAAAL13759-06
CAAAAL13759-22
CAAAAL13759-36
Isophorone diisocyanate (mixture of isomers) 98%
Isophorone diisocyanate
Purity: 98%
CAS No.: 4098-71-9
Molecular Formula: C12H18N2O2
Formula Weight: 222.29
Storage Temperature: 2°C to 8°C
Flash Point: 163°(325°F)
Refractive Index: 1.4840
Melting Point: -60°
Boiling Point: 153°/10mm
Beilstein Reference: 2726467
UN No.: 2290
MDL No.: MFCD00064956
Formula:
C₁₂H₁₈N₂O₂ Boiling Pt: 153 °C/10 mm Melting Pt: –60 °C Density: 1.061 Flash Pt: 163 °C(325 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00064956 CAS Number: 4098-71-9 EINECS: 223-861-6 UN: 2290 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...