5-Fluoro-2-toluoyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
5-Fluoro-2-methylbenzene-1-carbonyl chloride, 5-Fluoro-2-methylbenzoyl chloride
L13750-03
L13750-06
CAAAAL13750-03EA
42.34
CAD
CAAAAL13750-03
CAAAAL13750-06
5-Fluoro-2-toluoyl chloride 98%
5-Fluoro-2-toluoyl chloride
Purity: 98%
CAS No.: 21900-39-0
Molecular Formula: C8H6ClFO
Formula Weight: 172.59
Refractive Index: 1.5320
Boiling Point: 108°/24mm
UN No.: 3265
MDL No.: MFCD00236681
Formula:
C₈H₆ClFO Boiling Pt: 108 °C/24 mm Density: 1.270 Storage Temperature: Ambient |
MDL Number:
MFCD00236681 CAS Number: 21900-39-0 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...