Magnesium sulfate hydrate, (dried basis) contains ca 1-2 mol water of hydration

Supplier: Thermo Scientific Chemicals

L13739-0E L13739-22 L13739-36
CAAAAL13739-0EEA 134.46 CAD
CAAAAL13739-0E CAAAAL13739-22 CAAAAL13739-36
Magnesium sulfate hydrate, (dried basis) contains ca 1-2 mol water of hydration
Magnesium sulphate hydrate

CAS No.: 22189-08-8 Molecular Formula: MgSO4.xH2O Formula Weight: 120.37(anhy) Merck Reference: 14,5691 MDL No.: MFCD00149787

Formula: MgSO₄·nH₂O
Density: 2.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 22189-08-8
EINECS: 231-298-2
Merck Index: 14,5691

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR