Petroleum spirit, 60…80 °C

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Petroleum ether, Petroleum benzine

L13734-0F L13734-AU
CAAAAL13734-0FEA 535.87 CAD
CAAAAL13734-0F CAAAAL13734-AU
Petroleum spirit, 60…80 °C
Petroleum spirit (8032-32-4)

CAS No.: 8032-32-4 Flash Point: -10°(14°F) Odor: Petroleum-like Boiling Point: 60-80° Merck Reference: 14,5489 UN No.: 1268 MDL No.: MFCD00081849

Boiling Pt: 60…80 °C
Density: 0.672
Flash Pt: –10 °C (14 °F)
Storage Temperature: Ambient
CAS Number: 8032-32-4
EINECS: 232-453-7
UN: 1268
ADR: 3,III
Merck Index: 14,5489

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR