Isoamyl alcohol mixture of isomers 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L13660-0F
L13660-AE
L13660-AP
CAAAAL13660-0FEA
259.12
CAD
CAAAAL13660-0F
CAAAAL13660-AE
CAAAAL13660-AP
Isoamyl alcohol mixture of isomers 99%
Isoamyl alcohol
Purity: 99%
CAS No.: 123-51-3
Molecular Formula: (CH3)2CHCH2CH2OH
Formula Weight: 88.15
Flash Point: 45°(113°F)
Refractive Index: 1.4060
Odor: Unpleasant
Melting Point: -117°
Boiling Point: 130-132°
Beilstein Reference: 1718835
Merck Reference: 14,5195
UN No.: 1105
MDL No.: MFCD00002934
Formula:
(CH₃)₂CHCH₂CH₂OH Boiling Pt: 130…132 °C Melting Pt: –117 °C Density: 0.809 Flash Pt: 45 °C (113 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002934 CAS Number: 123-51-3 EINECS: 204-633-5 UN: 1105 ADR: 3,III Merck Index: 14,5195 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...