2,4-Dibromothiophene 90+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L13591-06
L13591-14
CAAAAL13591-03EA
66.94
CAD
CAAAAL13591-03
CAAAAL13591-06
CAAAAL13591-14
2,4-Dibromothiophene 90+%
2,4-Dibromothiophene
Purity: 90+%
CAS No.: 3140-92-9
Molecular Formula: C4H2Br2S
Formula Weight: 241.94
Flash Point:
Formula:
C₄H₂Br₂S Boiling Pt: 215 °C Melting Pt: –30 °C Density: 2.160 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00043889 CAS Number: 3140-92-9 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...