3-Chloro-2-thenoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Chloro-2-thiophenecarboxylic acid, 3-Chlorothiophene-2-carboxylic acid

L13560-03 L13560-06
CAAAAL13560-03EA 66.36 CAD
CAAAAL13560-03 CAAAAL13560-06
3-Chloro-2-thenoic acid 99%
3-Chloro-2-thenoic acid

Purity: 99% CAS No.: 59337-89-2 Molecular Formula: C5H3ClO2S Formula Weight: 162.60 Melting Point: 185-187° Beilstein Reference: 121052

Formula: C₅H₃ClO₂S
MW: 162.6 g/mol
Melting Pt: 185…187 °C
Storage Temperature: Ambient
MDL Number: MFCD00043888
CAS Number: 59337-89-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR