1,2,4,5-Tetrabromobenzene 94%

Supplier: Thermo Scientific Chemicals
Warning

L13539-09 L13539-18
CAAAAL13539-09EA 54.04 CAD
CAAAAL13539-09 CAAAAL13539-18
1,2,4,5-Tetrabromobenzene 94%
1,2,4,5-Tetrabromobenzene

Purity: 94% CAS No.: 636-28-2 Molecular Formula: C6H2Br4 Formula Weight: 393.70 Melting Point: 175-180° Beilstein Reference: 1365830

Formula: C₆H₂Br₄
MW: 393.7 g/mol
Melting Pt: 175…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00000063
CAS Number: 636-28-2
EINECS: 211-253-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR