2,5-Dichloroanisole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,4-Dichloro-2-methoxybenzene

L13497-06 L13497-14
CAAAAL13497-06EA 73.47 CAD
CAAAAL13497-06 CAAAAL13497-14
2,5-Dichloroanisole 98%
2,5-Dichloroanisole

Purity: 98% CAS No.: 1984-58-3 Molecular Formula: C7H6Cl2O Formula Weight: 177.03 Refractive Index: 1.5630 Boiling Point: 128-132°/3.5mm MDL No.: MFCD00061123

Formula: C₇H₆Cl₂O
MW: 177.03 g/mol
Boiling Pt: 128…132 °C (3.5 mmHg)
Melting Pt: 24
Density: 1.330
Storage Temperature: Ambient
MDL Number: MFCD00061123
CAS Number: 1984-58-3
EINECS: 217-852-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR