2-Chloroanilinium chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L13483-14
L13483-22
CAAAAL13483-14EA
66.04
CAD
CAAAAL13483-14
CAAAAL13483-22
2-Chloroanilinium chloride 97%
2-Chloroanilinium chloride
Purity: 97%
CAS No.: 137-04-2
Molecular Formula: C6H6ClN.HCl
Formula Weight: 164.03
Odor: Characteristic
Melting Point: ca 220° subl.
Beilstein Reference: 3624280
UN No.: 2018
MDL No.: MFCD00053282
Formula:
C₆H₇Cl₂N Melting Pt: ca 220 °C (subl.) Storage Temperature: Ambient |
MDL Number:
MFCD00053282 CAS Number: 137-04-2 EINECS: 205-274-7 UN: 2018 ADR: "6.1",II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...