3,4-Dicyanoaniline 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3,4-Dicyanoaniline, 4-Aminophthalonitrile
L13433-03
L13433-06
CAAAAL13433-03EA
79
CAD
CAAAAL13433-03
CAAAAL13433-06
3,4-Dicyanoaniline 98%
3,4-Dicyanoaniline
Purity: 98%
CAS No.: 56765-79-8
Molecular Formula: C8H5N3
Formula Weight: 143.15
Melting Point: 179-182°
Beilstein Reference: 2937382
UN No.: 3439
MDL No.: MFCD00041741
Formula:
C₈H₅N₃ MW: 143.15 g/mol Boiling Pt: 321 °C (1013 hPa) Melting Pt: 179…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041741 CAS Number: 56765-79-8 EINECS: 260-370-6 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...