6-Fluoro-m-toluidine 98%

Supplier: Thermo Scientific Chemicals
Danger

L13411-06 L13411-14
CAAAAL13411-06EA 38.24 CAD
CAAAAL13411-06 CAAAAL13411-14
6-Fluoro-m-toluidine 98%
6-Fluoro-m-toluidine

Purity: 98% CAS No.: 452-84-6 Molecular Formula: C7H8FN Formula Weight: 125.15 Flash Point: 81°(177°F) Refractive Index: 1.5330 Odor: Characteristic Boiling Point: 80-86°/11mm Beilstein Reference: 2715995 UN No.: 2810 MDL No.: MFCD00007654

Formula: C₇H₈FN
Boiling Pt: 80…86 °C/11 mm
Density: 1.109
Flash Pt: 81 °C (177 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007654
CAS Number: 452-84-6
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR