3-Methylpiperidine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L13408-14
L13408-22
CAAAAL13408-14EA
55.62
CAD
CAAAAL13408-14
CAAAAL13408-22
3-Methylpiperidine 98%
3-Methylpiperidine
Purity: 98%
CAS No.: 626-56-2
Molecular Formula: C6H13N
Formula Weight: 99.18
Flash Point: 8°(46°F)
Refractive Index: 1.4470
Odor: Amine-like
Melting Point: -24°
Boiling Point: 124-125°
Beilstein Reference: 79807
UN No.: 2733
MDL No.: MFCD00005994
Formula:
C₆H₁₃N MW: 99.18 g/mol Boiling Pt: 124…125 °C Melting Pt: –24 °C Density: 0.845 Flash Pt: 8 °C (46 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005994 CAS Number: 626-56-2 EINECS: 210-953-6 UN: 2733 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...